D-Fructose-1,6-bisphosphate trisodium salt octahydrate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium (3S,4R,5R)-6-((hydrogenphosphonato)oxy)-3,4,5-trihydroxy-2-oxohexyl phosphate octahydrate
A424452-100G
A424452-10G
A424452-25G
A424452-5G
76979-272EA
25.47
USD
76979-272
76979-274
76979-276
76979-278
D-Fructose-1,6-bisphosphate trisodium salt octahydrate 95%
D-Fructose-1,6-bisphosphate trisodium salt octahydrate
Precaución: for Research Use Only
Formula:
C₆H₂₇Na₃O₂₀P₂ MW: 550.179 g/mol |
MDL Number:
MFCD00149340 CAS Number: 81028-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...