3-(Bromomethyl)benzo[d]isoxazole 95%

Supplier: Ambeed
Danger

A466071-100MG A466071-1G A466071-250MG
76979-606EA 114.58 USD
76979-606 76979-608 76979-610
3-(Bromomethyl)benzo[d]isoxazole 95%
3-(Bromomethyl)benzo[d]isoxazole

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.04 g/mol
MDL Number: MFCD03701148
CAS Number: 37924-85-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR