Ethyl 4-bromobut-2-enoate 90%

Supplier: Ambeed
Danger

A881293-1G A881293-5G A881293-25G
76981-882EA 8.49 USD
76981-882 76981-884 77552-656
Ethyl 4-bromobut-2-enoate 90%
Ethyl 4-bromobut-2-enoate

Precaución: for Research Use Only

Formula: C₆H₉BrO₂
CAS Number: 6065-32-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR