N,N-Dimethylpyrrolidin-3-amine dihydrochloride 96%

Supplier: Ambeed
Warning

A139988-1G A139988-250MG A139988-25G A139988-5G
76972-544EA 37 USD
76972-544 76972-546 76972-548 76972-550
N,N-Dimethylpyrrolidin-3-amine dihydrochloride 96%
N,N-Dimethylpyrrolidin-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MDL Number: MFCD13561578
CAS Number: 50534-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR