Methyl-3,4-difluorobenzoate 97%

Supplier: Ambeed
Warning

A204789-100G A204789-1G A204789-25G A204789-500G A204789-5G
76975-320EA 43 USD
76975-320 76975-322 76975-324 76975-326 76975-328
Methyl-3,4-difluorobenzoate 97%
Methyl-3,4-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD00276597
CAS Number: 369-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR