(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 97%

Supplier: Ambeed
Warning

Synonyms: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

A444605-1G A444605-1MG A444605-25G A444605-5G A444605-100G
76979-414EA 18 USD
76979-414 76979-416 76979-418 76979-420 77673-092
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) 97%
(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₈
MDL Number: MFCD00063255
CAS Number: 369-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR