α,α,α-Trifluoro-m-toluonitrile 98%

Supplier: Ambeed
Danger

A396749-100G A396749-10G A396749-5G A396749-25G
76979-000EA 45 USD
76979-000 76979-002 76979-004 77568-806
α,α,α-Trifluoro-m-toluonitrile 98%
α,α,α-Trifluoro-m-toluonitrile

Precaución: For research use only.

Formula: C₈H₄F₃N
MDL Number: MFCD00001807
CAS Number: 368-77-4
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR