4,4'-Difluorobenzhydrol 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Difluorophenyl phenyl carbinol, 4,4'-Difluorobenzhydryl alcohol, 4,4'-Difluoro-α-phenylbenzyl alcohol, Bis(4-fluorophenyl)carbinol, 4,4'-Difluorodiphenylmethanol, Bis(4-fluorophenyl)methanol

A161286-100G A161286-10G A161286-25G A161286-5G A161286-500G
76973-606EA 35.01 USD
76973-606 76973-608 76973-610 76973-612 77600-288
4,4'-Difluorobenzhydrol 98%
4,4'-Difluorobenzhydrol

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
MW: 220.22 g/mol
MDL Number: MFCD00000357
CAS Number: 365-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR