Piroxicam 98%

Supplier: Ambeed
Warning

A265438-100G A265438-10G A265438-1G A265438-25G A265438-500G A265438-50MG A265438-5G
76977-088EA 68.99 USD
76977-088 76977-090 76977-092 76977-094 76977-096 76977-098 76977-100
Piroxicam 98%
Piroxicam

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₃O₄S
MDL Number: MFCD00057317
CAS Number: 36322-90-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR