N-Methyl-4-piperidinecarboxamide hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N-Methylpiperidine-4-carboxamide hydrochloride

A633174-100MG A633174-1G
76980-684EA 46.68 USD
76980-684 76980-686
N-Methyl-4-piperidinecarboxamide hydrochloride 95%
N-Methyl-4-piperidinecarboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD11615742
CAS Number: 1903-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR