2-Chloro-5-fluoro-4-methylpyrimidine 98%

Supplier: Ambeed
Warning

A229311-250MG A229311-100MG A229311-1G
76976-086EA 97 USD
76976-086 77600-836 77881-591
2-Chloro-5-fluoro-4-methylpyrimidine 98%
2-Chloro-5-fluoro-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂
MDL Number: MFCD14584604
CAS Number: 134000-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR