2-Bromo-6-methylbenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A570274-100MG A570274-1G A570274-250MG A570274-5G
76980-318EA 5 USD
76980-318 76980-320 76980-322 76980-324
2-Bromo-6-methylbenzo[d]thiazole 95%
2-Bromo-6-methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₆BrNS
MW: 228.10 g/mol
MDL Number: MFCD07783784
CAS Number: 3622-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR