4-Hydroxy-2-cyclopentenone 98%

Supplier: Ambeed
Warning

A186756-5G A186756-1G A186756-100MG A186756-250MG
76974-684EA 121 USD
76974-684 77567-720 77671-640 77671-642
4-Hydroxy-2-cyclopentenone 98%
4-Hydroxy-2-cyclopentenone

Precaución: For research use only.

Formula: C₅H₆O₂
MW: 98.09 g/mol
MDL Number: MFCD09035055
CAS Number: 61305-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR