1-Bromo-2,5-difluoro-3-nitrobenzene 97%

Supplier: Ambeed
Warning

A117497-100G A117497-10G A117497-1G A117497-25G A117497-5G
76971-294EA 1334.62 USD
76971-294 76971-296 76971-298 76971-300 76971-302
1-Bromo-2,5-difluoro-3-nitrobenzene 97%
1-Bromo-2,5-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MW: 237.986 g/mol
MDL Number: MFCD18205960
CAS Number: 741721-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR