2,7-Dichlorothiazolo[5,4-c]pyridine 95%

Supplier: Ambeed
Warning

A165213-100MG A165213-1G
76973-756EA 43.5 USD
76973-756 77671-326
2,7-Dichlorothiazolo[5,4-c]pyridine 95%
2,7-Dichlorothiazolo[5,4-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂S
MW: 205.0600 g/mol
MDL Number: MFCD13193435
CAS Number: 884860-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR