L-Tryptophan benzyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Trp-OBzl·HCl, H-Trp-OBzl.HCl

A281952-1G A281952-25G A281952-5G A281952-10G
76977-436EA 6.37 USD
76977-436 76977-438 76977-440 77672-454
L-Tryptophan benzyl ester hydrochloride 97%
L-Tryptophan benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₁₉ClN₂O₂
MW: 330.812 g/mol
MDL Number: MFCD00058122
CAS Number: 35858-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR