Methyl-4-(piperidin-4-yloxy)benzoate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(piperidin-4-yloxy)benzoate hydrochloride

A375256-100MG A375256-250MG
76978-762EA 47.75 USD
76978-762 76978-764
Methyl-4-(piperidin-4-yloxy)benzoate hydrochloride 98%
Methyl-4-(piperidin-4-yloxy)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₈ClNO₃
MW: 271.74 g/mol
MDL Number: MFCD12546302
CAS Number: 455323-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR