Piperidin-4-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A185399-100G A185399-1G A185399-25G A185399-5G
76974-592EA 389.36 USD
76974-592 76974-594 76974-596 76974-598
Piperidin-4-amine dihydrochloride 97%
Piperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
MDL Number: MFCD01570891
CAS Number: 35621-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR