(3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl) methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid tert-butyl ester 97%

Supplier: Ambeed
Warning

A196065-1G A196065-250MG A196065-25G A196065-5G
76975-040EA 47.75 USD
76975-040 76975-042 76975-044 76975-046
(3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl) methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid tert-butyl ester 97%
(3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl) methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₂₆H₃₆FN₃O₆S
MW: 537.64 g/mol
MDL Number: MFCD08460212
CAS Number: 355806-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR