Methyl-3-hydroxy-4-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
A227841-1G
A227841-250MG
A227841-5G
76975-996EA
12
USD
76975-996
76975-998
76976-000
Methyl-3-hydroxy-4-methylbenzoate 98%
Methyl-3-hydroxy-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ |
MDL Number:
MFCD00213808 CAS Number: 3556-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...