(1-Methylpyrrolidin-2-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyrrolidine-2-methanol, (S)-(-)-2-Hydroxymethyl-1-methylpyrrolidine, 1-METHYL-2-PYRROLIDINEMETHANOL

A199057-100G A199057-10G A199057-1G A199057-25G A199057-5G
76975-178EA 326.76 USD
76975-178 76975-180 76975-182 76975-184 76975-186
(1-Methylpyrrolidin-2-yl)methanol 97%
(1-Methylpyrrolidin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.176 g/mol
MDL Number: MFCD00011727
CAS Number: 3554-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR