3-((2,4-Dichlorobenzyl)oxy)pyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A116181-100MG A116181-250MG
76971-186EA 43.5 USD
76971-186 76971-188
3-((2,4-Dichlorobenzyl)oxy)pyrrolidine hydrochloride 95%
3-((2,4-Dichlorobenzyl)oxy)pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₃NO
MW: 282.59 g/mol
MDL Number: MFCD13561014
CAS Number: 1220033-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR