Methyl-2-(4-(tert-butyl)phenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-(tert-butyl)phenyl)acetate

A174608-100G A174608-10G A174608-500G A174608-5G A174608-25G
76974-186EA 65 USD
76974-186 76974-188 76974-190 76974-192 77082-558
Methyl-2-(4-(tert-butyl)phenyl)acetate 98%
Methyl-2-(4-(tert-butyl)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MDL Number: MFCD00051913
CAS Number: 3549-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR