2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Flecainide Impurity D

A326349-100G A326349-10G A326349-5G A326349-25G
76978-174EA 44.56 USD
76978-174 76978-176 76978-178 77672-680
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid 98%
2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈F₆O₄
MW: 318.17 g/mol
MDL Number: MFCD00221424
CAS Number: 35480-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR