N-(2-Hydroxypropyl)benzenesulfonamide 97%

Supplier: Ambeed
Warning

A111339-100G A111339-10G A111339-1G A111339-25G A111339-5G
76970-808EA 270.53 USD
76970-808 76970-810 76970-812 76970-814 76970-816
N-(2-Hydroxypropyl)benzenesulfonamide 97%
N-(2-Hydroxypropyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₃NO₃S
MW: 215.26 g/mol
MDL Number: MFCD00592915
CAS Number: 35325-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR