(R)-(-)-2-Amino-1-propanol 98%, ee 95%

Supplier: Ambeed
Danger

Synonyms: D-Alaninol, D(-)-2-Amino-1-propanol, D(-)-Alaninol, (R)-2-Aminopropan-1-ol

A140304-10G A140304-25G A140304-5G A140304-100G
76972-566EA 13 USD
76972-566 76972-568 76972-570 77592-664
(R)-(-)-2-Amino-1-propanol 98%, ee 95%
(R)-(-)-2-Amino-1-propanol

Precaución: for Research Use Only

Formula: C₃H₉NO
MDL Number: MFCD00064413
CAS Number: 35320-23-1
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR