Fmoc-3-methyl-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(3-Me)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(m-tolyl)propanoic acid

A120283-250MG A120283-5G
76971-466EA 18.04 USD
76971-466 76971-468
Fmoc-3-methyl-D-phenylalanine 97%
Fmoc-3-methyl-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD01863049
CAS Number: 352351-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR