2'-Hydroxy-3-phenylpropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxyphenyl)-3-phenylpropan-1-one, 2′-Hydroxy-3-phenylpropiophenone

A527232-500G A527232-100G A527232-25G A527232-1000G
76980-048EA 90.18 USD
76980-048 77602-246 77602-248 77673-398
2'-Hydroxy-3-phenylpropiophenone 98%
2'-Hydroxy-3-phenylpropiophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.2705 g/mol
MDL Number: MFCD00002221
CAS Number: 3516-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR