(R)-tert-Butyl-2-carbamoylpyrrolidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-tert-Butyl 2-carbamoylpyrrolidine-1-carboxylate, 1-Boc-D-prolinamide
A187778-100G
A187778-10G
A187778-1G
A187778-25G
A187778-5G
76974-722EA
254.62
USD
76974-722
76974-724
76974-726
76974-728
76974-730
(R)-tert-Butyl-2-carbamoylpyrrolidine-1-carboxylate 97%
(R)-tert-Butyl-2-carbamoylpyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈N₂O₃ MW: 214.27 g/mol |
MDL Number:
MFCD03419274 CAS Number: 70138-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...