1-(Triphenylmethyl)-L-histidine 97%

Supplier: Ambeed
Warning

Synonyms: H-His(Trt)-OH, 1-Trityl-L-histidine, H-His(1-Trt)-OH

A309637-25G A309637-5G A309637-100G A309637-10G A309637-500G
76977-874EA 19 USD
76977-874 76977-876 77601-398 77672-572 77672-574
1-(Triphenylmethyl)-L-histidine 97%
1-(Triphenylmethyl)-L-histidine

Precaución: for Research Use Only

Formula: C₂₅H₂₃N₃O₂
MDL Number: MFCD00153444
CAS Number: 35146-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR