2-Chloro-6-methylbenzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-methyl-1,3-benzothiazole, 2-Chloro-6-methylbenzo[d]thiazole

A507307-1G A507307-250MG A507307-5G
76979-928EA 50.93 USD
76979-928 76979-930 76979-932
2-Chloro-6-methylbenzothiazole 98%
2-Chloro-6-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS
MW: 183.66 g/mol
MDL Number: MFCD04448821
CAS Number: 3507-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR