4-(Trifluoromethoxy)phenyl isocyanate 98%

Supplier: Ambeed
Danger

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

A582063-1G A582063-25G A582063-5G
76980-388EA 5.31 USD
76980-388 76980-390 76980-392
4-(Trifluoromethoxy)phenyl isocyanate 98%
4-(Trifluoromethoxy)phenyl isocyanate

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₂
MW: 203.12 g/mol
MDL Number: MFCD00035702
CAS Number: 35037-73-1
UN: 2206
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR