3'-(Trifluoromethyl)acetophenone 98%
Supplier: Ambeed
|
Warning
|
A116660-100G
A116660-10G
A116660-500G
A116660-5G
A116660-25G
76971-234EA
35
USD
76971-234
76971-236
76971-238
76971-240
77566-118
3'-(Trifluoromethyl)acetophenone 98%
3'-(Trifluoromethyl)acetophenone
Precaución: For research use only.
Formula:
C₉H₇F₃O |
MDL Number:
MFCD00000391 CAS Number: 349-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...