3-Quinolinecarbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Cyanoquinoline

A334225-100MG A334225-1G A334225-250MG A334225-5G
76978-304EA 8.49 USD
76978-304 76978-306 76978-308 76978-310
3-Quinolinecarbonitrile 97%
3-Quinolinecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.17 g/mol
MDL Number: MFCD00006766
CAS Number: 34846-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR