4-Benzyl-N-carbobenzoxy-L-aspartate 98%

Supplier: Ambeed
Warning

A134496-10G A134496-100G A134496-25G A134496-500G
76972-284EA 13 USD
76972-284 76972-286 77599-938 77599-940 77881-065
4-Benzyl-N-carbobenzoxy-L-aspartate 98%
4-Benzyl-N-carbobenzoxy-L-aspartate

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₆
MDL Number: MFCD00037820
CAS Number: 3479-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR