Pentafluoro-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(perfluorophenyl)propanoic acid

A142712-100MG A142712-1G A142712-250MG A142712-5G
76972-672EA 22.28 USD
76972-672 76972-674 76972-676 77671-032
Pentafluoro-L-phenylalanine 95%
Pentafluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₆F₅NO₂
MW: 255.14 g/mol
MDL Number: MFCD01860881
CAS Number: 34702-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR