2,5-Dibromo-4-picoline 98%

Supplier: Ambeed
Warning

A159666-100G A159666-10G A159666-1G A159666-25G A159666-5G A159666-1000G A159666-500G
76973-476EA 47 USD
76973-476 76973-478 76973-480 76973-482 76973-484 77671-264 77671-266
2,5-Dibromo-4-picoline 98%
2,5-Dibromo-4-picoline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MDL Number: MFCD00234955
CAS Number: 3430-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR