(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Benzyloxycarbonyl-L-prolinamide, N-Cbz-L-prolinamide, N-Benzyloxycarbonyl-L-prolinamide, Benzyl (S)-2-carbamoylpyrrolidine-1-carboxylate, Z-Pro-NH₂
A109191-100G
A109191-10G
A109191-1G
A109191-25G
A109191-5G
76970-610EA
90.18
USD
76970-610
76970-612
76970-614
76970-616
76970-618
(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate 95%
(S)-Benzyl-2-carbamoylpyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₆N₂O₃ MW: 248.28 g/mol |
MDL Number:
MFCD00020830 CAS Number: 34079-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...