1,3-Bis(4,5-dihydro-2-oxazolyl)benzene 95%

Supplier: Ambeed
Warning

A132823-100G A132823-10G A132823-25G A132823-500G
76972-180EA 59.42 USD
76972-180 76972-182 76972-184 76972-186
1,3-Bis(4,5-dihydro-2-oxazolyl)benzene 95%
1,3-Bis(4,5-dihydro-2-oxazolyl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD00191606
CAS Number: 34052-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR