3,4-Difluorobenzaldehyde 96%

Supplier: Ambeed
Warning

A227855-100G A227855-10G A227855-25G A227855-500G A227855-5G
76976-002EA 78.51 USD
76976-002 76976-004 76976-006 76976-008 76976-010
3,4-Difluorobenzaldehyde 96%
3,4-Difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O
MW: 142.1 g/mol
MDL Number: MFCD00010328
CAS Number: 34036-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR