4-Chloro-D-phenylalanine methyl ester hydrochloride 97%

Supplier: Ambeed
Warning

A178960-250MG A178960-5G A178960-1G
76974-356EA 18.04 USD
76974-356 76974-358 77600-450
4-Chloro-D-phenylalanine methyl ester hydrochloride 97%
4-Chloro-D-phenylalanine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂·HCl
MDL Number: MFCD00236920
CAS Number: 33965-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR