Succinimido (S)-2-[(tert-butoxycarbonyl)amino]propionate 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-alanine N-succinimidyl ester, Boc-Ala-OSu

A140924-25G A140924-5G A140924-100G
76972-614EA 46 USD
76972-614 76972-616 77881-101
Succinimido (S)-2-[(tert-butoxycarbonyl)amino]propionate 95%
Succinimido (S)-2-[(tert-butoxycarbonyl)amino]propionate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₆
MDL Number: MFCD00065974
CAS Number: 3392-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR