6-Chloro-N-methyl-3-nitropyridin-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-nitro-6-methylaminopyridine

A133552-100MG A133552-1G A133552-250MG A133552-5G A133552-25G
76972-218EA 9 USD
76972-218 76972-220 76972-222 76972-224 77881-061
6-Chloro-N-methyl-3-nitropyridin-2-amine 95%
6-Chloro-N-methyl-3-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O₂
MDL Number: MFCD08460164
CAS Number: 33742-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR