Disodium-4-formylbenzene-1,3-disulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Formylbenzene-1,3-disulphonic acid disodium salt, Sodium 4-formylbenzene-1,3-disulphonate

A158430-500G A158430-5G A158430-100G
76973-424EA 96 USD
76973-424 76973-426 77881-237
Disodium-4-formylbenzene-1,3-disulfonate 98%
Disodium-4-formylbenzene-1,3-disulfonate

Precaución: for Research Use Only

Formula: C₇H₆O₇S₂
MDL Number: MFCD00043913
CAS Number: 33513-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR