2-Amino-5-bromo-3-(hydroxymethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-bromo-3-pyridinemethanol

A146093-10G A146093-1G A146093-250MG
76972-844EA 127.31 USD
76972-844 76972-846 76972-848
2-Amino-5-bromo-3-(hydroxymethyl)pyridine 98%
2-Amino-5-bromo-3-(hydroxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD05664025
CAS Number: 335031-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR