2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%

Supplier: Ambeed
Warning

Synonyms: Disodium 2'-deoxy-5'-O-phosphonatoguanosine, 2'-Deoxyguanosine-5'-monophosphoric acid disodium salt

A463091-100MG A463091-1G A463091-250MG A463091-5G
76979-566EA 33.95 USD
76979-566 77602-014 77673-182 77673-184
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₅Na₂O₇P·xH₂O
MW: 391.18 g/mol (anhy)
MDL Number: MFCD00080331
CAS Number: 33430-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR