4-Piperidinyl cyclopropanecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A362727-1G A362727-250MG A362727-5G
76978-606EA 47 USD
76978-606 76978-608 76978-610
4-Piperidinyl cyclopropanecarboxylate hydrochloride 95%
4-Piperidinyl cyclopropanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₆ClNO₂
MDL Number: MFCD13559803
CAS Number: 1219968-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR