2-(6-Chloro-1H-benzo[d][1,2,3]triazol-1-yl)-1,1,3,3-tetramethyluronium hexafluorophosphate(V) 97%

Supplier: Ambeed
Danger

A138566-10G A138566-25G
76972-472EA 9.55 USD
76972-472 76972-474
2-(6-Chloro-1H-benzo[d][1,2,3]triazol-1-yl)-1,1,3,3-tetramethyluronium hexafluorophosphate(V) 97%
2-(6-Chloro-1H-benzo[d][1,2,3]triazol-1-yl)-1,1,3,3-tetramethyluronium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClF₆N₅OP
MW: 413.69 g/mol
MDL Number: MFCD04973268
CAS Number: 330645-87-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR