1-(Bis(dimethylamino)methylene)-5-chloro-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide tetrafluoroborate 97%

Supplier: Ambeed
Danger

A200578-100G A200578-10G A200578-25G A200578-5G
76975-236EA 77 USD
76975-236 76975-238 76975-240 76975-242
1-(Bis(dimethylamino)methylene)-5-chloro-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide tetrafluoroborate 97%
1-(Bis(dimethylamino)methylene)-5-chloro-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClF₄N₅O
MW: 355.53 g/mol
MDL Number: MFCD08461912
CAS Number: 330641-16-2
UN: 2921
ADR: 8,4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR