5-Ethoxypyridin-2-amine 98%

Supplier: Ambeed
Warning

A135543-250MG A135543-5G A135543-1G A135543-100MG
76972-330EA 29 USD
76972-330 76972-332 77611-218 77881-074
5-Ethoxypyridin-2-amine 98%
5-Ethoxypyridin-2-amine

Precaución: for Research Use Only

Formula: C7H10N2O
MDL Number: MFCD11135752
CAS Number: 89943-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR